Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mss51
Entrez Gene ID
289904
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
Zmynd17
Chromosome Location
Chr.15: 4240203 - 4252072 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106025.1 NP_001099495.1 1 - 2 50 71

View Full Product Details