Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vnn1
Entrez Gene ID
29142
Gene Name
vanin 1
Gene Aliases
Tiff66
Chromosome Location
Chr.1: 22614832 - 22625134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025623.1 NP_001020794.1 2 - 3 480 79
GenBank mRNA BC098934.1 - 2 - 3 480 79

View Full Product Details