Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pter
Entrez Gene ID
63852
Gene Name
phosphotriesterase related
Gene Aliases
Rpr-1, Rpr1
Chromosome Location
Chr.17: 80250443 - 80311987 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022224.2 NP_071560.2 2 - 3 494 98
XM_006254290.2 XP_006254352.2 2 - 3 3763 98
XM_006254291.2 XP_006254353.1 2 - 3 581 98
GenBank mRNA BC078833.1 - 2 - 3 494 98

View Full Product Details