Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eomes
Entrez Gene ID
316052
Gene Name
eomesodermin
Gene Aliases
-
Chromosome Location
Chr.8: 127144368 - 127152618 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766711.1 XP_008764933.1 6 - 7 2159 73
GenBank mRNA AY457971.1 - 3 - 4 433 73

View Full Product Details