Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apip
Entrez Gene ID
295961
Gene Name
APAF1 interacting protein
Gene Aliases
RGD1564562
Chromosome Location
Chr.3: 92969129 - 92998104 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106492.1 NP_001099962.1 3 - 4 316 68
XM_006234638.2 XP_006234700.1 3 - 4 365 68

View Full Product Details