The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009708.1 | NP_001009708.1 | - | 604 | 87 |
XM_006233438.2 | XP_006233500.1 | - | 370 | 87 | |
GenBank mRNA | BC087700.1 | - | - | 604 | 87 |
FQ233103.1 | - | - | 447 | 87 |