The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109676.2 | NP_001103146.1 | 2 - 3 | 694 | 107 |
GenBank mRNA | AJ518866.1 | - | 2 - 3 | 694 | 107 |