Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntf3
Entrez Gene ID
81737
Gene Name
neurotrophin 3
Gene Aliases
-
Chromosome Location
Chr.4: 158636883 - 158705964 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763294.1 XP_008761516.1 1 - 2 269 123

View Full Product Details