Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Emilin3
Entrez Gene ID
362262
Gene Name
elastin microfibril interfacer 3
Gene Aliases
-
Chromosome Location
Chr.3: 156899892 - 156906566 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109901.2 NP_001103371.1 1 - 2 312 102
XM_006235495.1 XP_006235557.1 1 - 2 986 102

View Full Product Details