Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudcd3
Entrez Gene ID
100125378
Gene Name
NudC domain containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 86326413 - 86333397 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103362.1 NP_001096832.1 - 530 83
GenBank mRNA BC107925.1 - - 530 83

View Full Product Details