The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103362.1 | NP_001096832.1 | - | 655 | 59 |
GenBank mRNA | BC107925.1 | - | - | 655 | 59 |