Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sun5
Entrez Gene ID
296289
Gene Name
Sad1 and UNC84 domain containing 5
Gene Aliases
Spag4l
Chromosome Location
Chr.3: 149336114 - 149356166 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106530.1 NP_001100000.1 10 - 11 900 69
XM_008762312.1 XP_008760534.1 12 - 13 1071 69

View Full Product Details