Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb20
Entrez Gene ID
641641
Gene Name
defensin beta 20
Gene Aliases
-
Chromosome Location
Chr.3: 147952879 - 147955502 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037517.1 NP_001032606.1 1 - 2 47 81
GenBank mRNA AY621351.1 - 1 - 2 47 81

View Full Product Details