The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031571.2 | NP_113759.1 | 9 - 10 | 1231 | 66 |
XM_008761703.1 | XP_008759925.1 | 10 - 11 | 1310 | 66 | |
GenBank mRNA | S48190.1 | - | 9 - 10 | 1231 | 66 |