The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128137.1 | NP_001121609.1 | 4 - 5 | 442 | 68 |
GenBank mRNA | DQ537494.1 | - | 4 - 5 | 442 | 68 |