Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgfbp3
Entrez Gene ID
499349
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
RGD1560326
Chromosome Location
Chr.1: 255555050 - 255557055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109165.1 NP_001102635.1 1 - 2 78 70

View Full Product Details