Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vasn
Entrez Gene ID
679921
Gene Name
vasorin
Gene Aliases
-
Chromosome Location
Chr.10: 11121041 - 11131548 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109382.1 NP_001102852.1 1 - 2 139 91

View Full Product Details