Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc22
Entrez Gene ID
298300
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
RGD1561518
Chromosome Location
Chr.5: 126225529 - 126243178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106671.1 NP_001100141.1 4 - 5 959 79

View Full Product Details