The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053397.1 | NP_445849.1 | 3 - 4 | 315 | 85 |
XM_006238684.2 | XP_006238746.1 | 1 - 2 | 3829 | 85 | |
XM_006238685.2 | XP_006238747.1 | 2 - 3 | 3610 | 85 | |
XM_006238687.2 | XP_006238749.1 | 2 - 3 | 3565 | 85 | |
XM_006238691.2 | XP_006238753.1 | 1 - 2 | 3829 | 85 | |
GenBank mRNA | AY230412.1 | - | 1 - 2 | 58 | 85 |
BC079078.1 | - | 3 - 4 | 315 | 85 |