The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013212.1 | NP_001013230.1 | 8 - 9 | 1096 | 92 |
GenBank mRNA | BC091186.1 | - | 8 - 9 | 1096 | 92 |