Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eefsec
Entrez Gene ID
500255
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA-specific
Gene Aliases
RGD1560552
Chromosome Location
Chr.4: 120194335 - 120390202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109249.1 NP_001102719.1 5 - 6 948 73
XM_006236864.2 XP_006236926.1 6 - 7 1622 73
GenBank mRNA BC168694.1 - 6 - 7 1585 73

View Full Product Details