Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hif1an
Entrez Gene ID
309434
Gene Name
hypoxia-inducible factor 1, alpha subunit inhibitor
Gene Aliases
-
Chromosome Location
Chr.1: 264309214 - 264319269 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113749.1 NP_001107221.1 3 - 4 595 100
GenBank mRNA BC158810.1 - 3 - 4 595 100

View Full Product Details