The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013905.1 | NP_001013927.1 | 5 - 6 | 523 | 75 |
XM_006231450.2 | XP_006231512.1 | 5 - 6 | 564 | 75 | |
GenBank mRNA | BC078988.1 | - | 5 - 6 | 523 | 75 |