Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Panx3
Entrez Gene ID
315567
Gene Name
pannexin 3
Gene Aliases
-
Chromosome Location
Chr.8: 40126379 - 40137390 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199398.1 NP_955430.1 2 - 3 626 67
GenBank mRNA AJ557017.1 - 2 - 3 626 67

View Full Product Details