The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106172.1 | NP_001099642.1 | - | 577 | 61 |
XM_008772370.1 | XP_008770592.1 | 3 - 4 | 1835 | 61 | |
XM_008772371.1 | XP_008770593.1 | 3 - 4 | 650 | 61 | |
XM_008772372.1 | XP_008770594.1 | 3 - 4 | 1576 | 61 | |
XM_008772373.1 | XP_008770595.1 | 3 - 4 | 627 | 61 |