Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcp10b
Entrez Gene ID
308169
Gene Name
t-complex protein 10b
Gene Aliases
-
Chromosome Location
Chr.1: 53531007 - 53554113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107461.1 NP_001100931.1 4 - 5 748 64
XM_006227927.2 XP_006227989.1 6 - 7 856 64
XM_006227928.2 XP_006227990.1 6 - 7 856 64
XM_006227930.2 XP_006227992.1 6 - 7 856 64
XM_008758757.1 XP_008756979.1 4 - 5 750 64
XM_008758758.1 XP_008756980.1 4 - 5 750 64
XM_008758759.1 XP_008756981.1 5 - 6 793 64
GenBank mRNA BC166440.1 - 6 - 7 787 64

View Full Product Details