Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-M6-2
Entrez Gene ID
365527
Gene Name
RT1 class I, locus M6, gene 2
Gene Aliases
-
Chromosome Location
Chr.20: 2075796 - 2079183 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772695.1 XP_008770917.1 1 - 2 1061 89

Gene Symbol
RT1-M6-1
Entrez Gene ID
414785
Gene Name
RT1 class I, locus M6, gene 1
Gene Aliases
-
Chromosome Location
Chr.20: 2057877 - 2060794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772698.1 XP_008770920.1 2 - 3 360 89
GenBank mRNA EU106176.1 - 1 - 2 274 89

View Full Product Details