Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc85a
Entrez Gene ID
289855
Gene Name
coiled-coil domain containing 85A
Gene Aliases
RGD1311700
Chromosome Location
Chr.14: 112719482 - 112900724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191553.1 NP_001178482.1 - 1427 66

View Full Product Details