Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scg2
Entrez Gene ID
24765
Gene Name
secretogranin II
Gene Aliases
Chcg
Chromosome Location
Chr.9: 85237460 - 85243033 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022669.1 NP_073160.1 1 - 1 1933 135
XM_006245148.2 XP_006245210.1 3 - 3 2003 135
XM_008767195.1 XP_008765417.1 2 - 2 2257 135
GenBank mRNA BC061549.1 - 3 - 3 1855 135
M93669.1 - 1 - 1 1933 135

View Full Product Details