Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-DOb
Entrez Gene ID
365542
Gene Name
RT1 class II, locus DOb
Gene Aliases
DOb, H2-Ob, RT1-Bb2
Chromosome Location
Chr.20: 4020183 - 4026398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008846.1 NP_001008846.1 5 - 6 784 71
XM_006255970.2 XP_006256032.1 5 - 6 908 71
GenBank mRNA KC222915.1 - 5 - 6 784 71
KC222916.1 - 5 - 6 784 71
KC222930.1 - 5 - 6 706 71
KC222931.1 - 5 - 6 706 71

View Full Product Details