Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rsl1d1
Entrez Gene ID
302898
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
Csig
Chromosome Location
Chr.10: 4348267 - 4502568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008876.2 NP_001008876.1 4 - 5 330 121
GenBank mRNA AY310144.1 - 4 - 5 330 121

Gene Symbol
NEWGENE_1359295
Entrez Gene ID
103693266
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
Rsl1d1
Chromosome Location
Chr.10: 4446764 - 4459120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767502.1 XP_008765724.1 - 903 121

View Full Product Details