The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080904.2 | NP_543180.1 | 3 - 4 | 461 | 119 |
XM_006257320.2 | XP_006257382.1 | 4 - 5 | 670 | 119 | |
GenBank mRNA | AY325223.1 | - | 4 - 5 | 428 | 119 |
BC088865.1 | - | 3 - 4 | 461 | 119 | |
L12382.1 | - | 4 - 5 | 574 | 119 |