The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008811.1 | NP_001008811.1 | 4 - 5 | 793 | 151 |
XM_006242351.2 | XP_006242413.1 | 4 - 5 | 849 | 151 | |
GenBank mRNA | AB355640.1 | - | 4 - 5 | 793 | 151 |