The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013145.1 | NP_001013163.1 | 2 - 2 | 1261 | 106 |
XM_006227922.2 | XP_006227984.1 | 3 - 3 | 1371 | 106 | |
XM_008758755.1 | XP_008756977.1 | 2 - 2 | 1138 | 106 | |
XM_008758756.1 | XP_008756978.1 | 2 - 2 | 3187 | 106 | |
GenBank mRNA | BC091197.1 | - | 2 - 2 | 1261 | 106 |
EF547659.1 | - | 1 - 1 | 996 | 106 | |
FQ225325.1 | - | 1 - 1 | 634 | 106 | |
FQ226017.1 | - | 2 - 2 | 1138 | 106 |