The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037529.1 | NP_001032618.1 | 1 - 2 | 50 | 75 |
XM_008770775.1 | XP_008768997.1 | 2 - 3 | 351 | 75 | |
GenBank mRNA | AY621371.1 | - | 1 - 2 | 50 | 75 |