The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085475.1 | NP_001078944.1 | 1 - 1 | 1347 | 153 |
GenBank mRNA | BC127531.1 | - | 1 - 1 | 1347 | 153 |