Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir17
- Entrez Gene ID
-
100313998
- Gene Name
- microRNA 17
- Gene Aliases
-
Mir17-1, rno-mir-17-1
- Chromosome Location
-
Chr.15: 100179879 - 100179962 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_031818.1 |
|
1 - 1 |
40 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SLIT and NTRK-like family, member 5 |
Slitrk5 |
|
-3.4mb
|
Sense |
Chr. - 96817892 - 96826760 on Build Rnor_6.0 |
Downstream |
microRNA 18a |
Mir18a |
rno-mir-18a |
53.0
|
Sense |
Chr. - 100180015 - 100180110 on Build Rnor_6.0 |
View Full Product Details