Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRS2
Entrez Gene ID
851207
Gene Name
Gene Aliases
FUN38, SWA3, YAL026C
Chromosome Location
Chr.I: 95630 - 99697 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178171.2 NP_009376.2 1 - 1 3948 74

View Full Product Details