Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLC2
Entrez Gene ID
851246
Gene Name
Gene Aliases
YAL053W
Chromosome Location
Chr.I: 45899 - 48250 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178196.2 NP_009348.2 1 - 1 2252 97

View Full Product Details