Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTH2
Entrez Gene ID
852223
Gene Name
Gene Aliases
YBL057C
Chromosome Location
Chr.II: 112801 - 113427 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178297.2 NP_009496.2 1 - 1 537 100

View Full Product Details