Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFT2
Entrez Gene ID
852172
Gene Name
Gene Aliases
YBL102W
Chromosome Location
Chr.II: 24098 - 24745 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178342.1 NP_009448.1 1 - 1 549 81

View Full Product Details