Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG22
Entrez Gene ID
850319
Gene Name
Gene Aliases
AUT4, YCL038C
Chromosome Location
Chr.III: 54941 - 56527 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178683.1 NP_009892.1 1 - 1 1448 91

View Full Product Details