Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL32
Entrez Gene ID
850359
Gene Name
Gene Aliases
YCR003W, bL32m
Chromosome Location
Chr.III: 118620 - 119171 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178716.1 NP_009929.1 1 - 1 446 116

View Full Product Details