Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPZ2
Entrez Gene ID
852046
Gene Name
Gene Aliases
YDR436W
Chromosome Location
Chr.IV: 1334821 - 1336953 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180744.1 NP_010724.1 1 - 1 1048 86

View Full Product Details