Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRS4
Entrez Gene ID
852049
Gene Name
Gene Aliases
YDR439W
Chromosome Location
Chr.IV: 1339676 - 1340719 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180747.1 NP_010727.1 1 - 1 954 70

View Full Product Details