Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUG1
Entrez Gene ID
856934
Gene Name
Gene Aliases
YER185W
Chromosome Location
Chr.V: 559454 - 560365 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179075.3 NP_011112.3 1 - 1 551 65

View Full Product Details