Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PES4
Entrez Gene ID
850579
Gene Name
Gene Aliases
YFR023W
Chromosome Location
Chr.VI: 199874 - 201709 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179988.1 NP_116678.1 1 - 1 1649 80

View Full Product Details