Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YHB1
Entrez Gene ID
853149
Gene Name
Gene Aliases
YGR234W, YHB4
Chromosome Location
Chr.VII: 959904 - 961103 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181363.1 NP_011750.1 1 - 1 444 97

View Full Product Details