Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIN1
Entrez Gene ID
856561
Gene Name
Gene Aliases
SNU40, YHR156C
Chromosome Location
Chr.VIII: 411384 - 412406 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179287.1 NP_012026.1 1 - 1 852 90

View Full Product Details