Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDR11
Entrez Gene ID
854802
Gene Name
Gene Aliases
YIL013C
Chromosome Location
Chr.IX: 328207 - 332442 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179363.1 NP_012252.1 1 - 1 3952 84

View Full Product Details